Gas Collecting Bulb, with Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
7395-04 7395-18 7395-30 7395-40 7395-44 7395-48 7395-50
89058-794EA 193.52 CAD
89058-794 89058-796 89058-798 89058-800 89058-802 89058-804 89058-806
Gas Collecting Bulb, with Ace-Thred, Ace Glass Incorporated
Samplers Gas Samplers
Designed with #7 Ace-Thred end valves that ensure a tight fit without grease
Operation is simple. With bushings at fingertip tightness, push plungers into bulb until flow holes are inside O-Rings (enlarged end of plunger acts as an automatic stop). Valves are now open and ready for flush sampling. When sampling is complete, and with bushing still at finger tip tightness, pull plungers out so flow holes are outside O-Rings. Valves are now closed. Bushings can then be tightened more for handling until ready to analyze.
A bulge is built on the small end of the plunger to help guardMore Product Information