Flask Heater, Ace Glass Incorporated
Supplier: Ace Glass
6494-10 6494-15 6494-20 6494-25
89054-996EA 1576.44 CAD
89054-996 89054-998 89055-000 89055-002
Flask Heater, Ace Glass Incorporated
Heaters Heating Mantles
Specially designed glass/fiber cloth mantle with bottom opening and zipper for use with flasks to allow valve to protrude below heater
Upper temperature limit is 450 °C (842 °F). Must be operated through a temperature controller.
Certifications: CSA approved.
Ordering information: Flask heater is supplied with 122 cm (4') cord and locking connector.