SP Bel-Art® Economy Dippers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36785-0016 F36784-0016 F36786-0016
47727-678EA 240.87 CAD
47727-678 47727-676 47727-680
SP Bel-Art® Economy Dippers, Bel-Art Products, a part of SP
Designed for convenient sampling from tanks, vats, streams, ponds, lakes, etc., these economy dippers are lightweight, easy to use, and portable.
  • Polypropylene cup construction
  • Economical
  • Ribbed for a sure grip
  • Polyethylene handles
  • End hook for convenient hanging storage
The polypropylene cups have threaded fittings that screw onto the handle. Two pouring spouts on the 500 mL (16oz.) dipper make it useful for left or right handed users to pour from either side. The high-density polyethylene handles are strong honeycomb tubes without inserts. They are ribbed for a sure grip. The end of the handle has a hook to facilitate hanging storage.