SP Bel-Art® Adjustable Handle Water Sample Dipper, Bel-Art Products, a part of SP
Supplier: Bel-Art
Scienceware®
F37807-0000
66175-065EA 433.35 CAD
66175-065
SP Bel-Art® Adjustable Handle Water Sample Dipper, Bel-Art Products, a part of SP
Samplers Sample Dippers
Designed for taking samples of water, sewage, wastes, and effluents from lakes, streams, tanks, or ponds, the water sample dipper provides a long reach for otherwise inaccessible spots.
  • Polypropylene construction
  • Adjustable aluminum handle
  • Suited to sampling aqueous solutions
  • Easy to adjust for optimal comfort
A 1000 mL graduated polypropylene pitcher is mounted onto an aluminum tube handle. The handle has three parts, a 91 cm (3') lower section, a 112 cm (44") telescoping section, and an additional 91 cm (3') upper section. Connections are made with spring buttons with adjusting steps, 15 cm (6") apart. Handle diameter is 2.5 cm (1") with 2.2 cm (⅞") telescoping tube.