Buffer Solutions in FIOLAX® Ampoules, SI Analytics
Supplier: SI Analytics
285138192
285138398
285138238
97042-532EA
962.27
CAD
97042-532
97042-554
97042-540
Buffer Solutions in FIOLAX® Ampoules, SI Analytics
Buffers
pH Calibration Buffers
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...