CritiCore Indura Unisex Flame Resistant Lab Coats
Supplier: VWR International
89426-882 89426-884 89426-886 89426-888 89426-890 89426-892
89426-882EA 235.5 CAD
89426-882 89426-884 89426-886 89426-888 89426-890 89426-892 89426-894 89426-896 89426-898
CritiCore Indura Unisex Flame Resistant Lab Coats
Workwear
CritiCore has developed flame-resistant lab coats to provide the protection required in many of today's high-tech laboratories and research facilities.
  • Flame-resistant identification patches sewn on the back and both sleeves for easy recognition
  • Quick-release snap front for fast removal
  • Safety-sleeve cuff with tunneled elastic
  • Two oversized waist pockets and one breast pocket with a convenient pencil slot
  • Vent openings on the sides allow access to pant pockets
These coats are made with 100% cotton fabric, durable enough to withstand industrial laundering while maintaining fire-resistant properties for the life of the garment.
Certifications: CAT I
Ordering information: Available only in blue. Coats in sizes X-Small to XX-Large are stocked. For sizes 3X-Large to 6X-Large, extended lead time may be required. Contact a Avantor sales representative for more information.