Immersion Coolers, FT-25, PolyScience
Supplier: VWR International
89400-958 89400-962 89400-964 89400-966 89400-968
89400-958EA 12490.77 CAD
89400-958 89400-962 89400-964 89400-966 89400-968
Immersion Coolers, FT-25, PolyScience
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Enhance and simplify your low-temperature laboratory work with these low temperature coolers. Available as immersion probe and flow-through models, these units provide rapid, low-cost cooling of liquids to temperatures as low as -100 °C. Typical applications include cooling exothermic reactions, freeze point determinations, freeze drying, impact testing, lyophilization and vapor and solvent trapping.
Flow-through style coolers offer an extremely economical alternative to tap water cooling. Use with heated circulating baths to reach temperatures below ambient. Use with refrigerated circulating baths to increase cooling capacity.
Immersion probe style coolers are available with different fixed temperatures. Excellent for trapping and Dewar-type applications, these coolers are designed to run at maximum cooling and reduce the expense of using dry ice or liquid nitrogen. A flexible hose allows convenient placement of the cooling probe.
Caution: Please note these units are all 240 V, 50 Hz.