Corning® ITS Universal Culture Supplement Premix, Corning
Supplier: Corning
354350
354351
354352
CACB354350EA
588.43
CAD
CACB354350
CACB354351
CACB354352
Corning® ITS Universal Culture Supplement Premix, Corning
Cell Culture Supplements and Additives
ITS contains insulin, human transferrin, and selenous acid, the three most universally essential components of defined culture media.
They stimulate cell proliferation of a variety of cells under serum-reduced conditions.
Tested for ability to promote proliferation of HeLa cells in serum-free medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.