BLItz™ Label-Free Protein Analysis System, ForteBio
Supplier: Forte Bio
18-5001 18-5082 18-5022 18-5101 18-5096 18-5019 18-5092 18-5088 18-5060 18-5045
CA89413-822EA 45116.66 CAD
CA89413-822 CA89413-824 CA89413-826 CA89413-828 CA89413-834 CA89413-836 CA89413-840 CA89413-842 CA89413-844 CA89413-846 CA89413-848 CA89413-850
BLItz™ Label-Free Protein Analysis System, ForteBio
Analysers Protein Analysers
The BLItz system for label-free assays is designed for benchtop protein analysis and characterization
The BLItz system uses ForteBio's Dip and Read™ label-free assays. These direct binding assays take place on a disposable biosensor made from a biocompatible matrix that is uniform, non-denaturing, and minimizes non-specific binding. Only molecules that bind directly to the biosensor surace are detected, providing high specificy, even in complex mixtures. A variety of disposable biosensors are available to run a wide range of assays.
Use only a 4 µl drop of sample for diverse assays such as monitoring binding interactions, quantifying expression, analyzing column fractions, and even developing optimized immunoassays. The BLItz is an affordably-priced system with a simplified workflow to determine rate and affinity constants in minutes. Able to detect analytes in lysates or media, the BLItz system detects the presence or absence of protein and measures concentrations in seconds to minutes, compared to much longer gel and blot-based methods.
Ordering information: System includes BLItz Pro™ software, two (2) drop holders, power cord, USB cable, and a Quick Start Guide.