Calibration Baths, PolyScience
Supplier: Polyscience
AP15RCAL-A11B PD15HCAL-A12E PP15HCAL-A11B PD15HCAL-A11B PP15HCAL-A12E
89400-936EA 10690.95 CAD
89400-936 89400-938 89400-940 89400-942 89400-944
Calibration Baths, PolyScience
Circulating Thermostats Open Circulator Systems Calibration Circulators
PolyScience Calibration Baths accommodate up to 6 temperature measuring devices and come with 2-point factory calibration, as well as bath lids with sleeves for two 3mm, 4mm and 6mm diameter openings
Calibration Baths are available in Heated and Refrigerated/Heated models. Swivel 180™, LidDock™ and DuraTop™ features all come standard. WhisperCool® Environmental Control System is standard on refrigerated units. All models include USB-A, RS-232, RS-485 and Ethernet, and include an external temperature probe port.
Event scheduling (time and date), along with real-time clock and temperature trends (for up to 10 days) included. Drain ports make it easy to remove liquids from the bath and the washable air filter helps keep the circulator energy-efficient and running at peak performance.
Available with the Performance Programmable (OpenMode programming for unlimited steps and programs, for specific test protocols) or Performance Digital Controller, all Calibration Baths feature an icon-driven SmartTouch interface, which leads you through menus, functions and options, enabling you to make any adjustments, even with your gloves on. All screens and menus, including on-screen help, can be displayed in any of 11 different languages.
Certifications: Complies with DIN 12876-1 Class III safety requirements for use with flammable liquids, cETLus compliant.