SP Bel-Art Stack Rack™ Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18860-2630 F18860-1013 F18860-1316 F18860-1620 F18860-2125
60917-462CS 503.69 CAD
60917-462 60917-450 60917-453 60917-456 60917-459
SP Bel-Art Stack Rack™ Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
These sturdy, rigid racks feature interlocking end plates for secure stacking when loaded with tubes up to 10 cm (4") long.
  • Polypropylene or polycarbonate construction
  • Sturdy, rigid designs
  • Decks may be white or clear depending on model
  • Numbered and lettered rows
  • Autoclavable
End plates have handles
The white surfaces of polypropylene racks make color comparisons easy. Sturdy and chemical-resistant, these racks will not shatter or stain. They are steam autoclavable at 121°C (250°F). Top shelf height is 7 cm (2³/₄").
Polycarbonate racks have crystal-clear decks and are perfect for submersing in water baths. Top shelf height is 7.3 cm (2⁷/₈"). Steam autoclavable at 121°C (250°F).
Rows are numbered and lettered for easy tube identification. Choose from polypropylene or polycarbonate to best suit your needs.