Anti-Transferrin Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
VAP-EN004D VAP-EN004F
CA95045-166EA 346.02 CAD
CA95045-166 CA95045-168
Anti-Transferrin Rabbit Polyclonal Antibody
Antibodies
Transferrin is a serum glycoprotein involved in iron transport. Transferrin exists in two forms, the iron-free Apotransferrin, which binds two Fe3+ ions to generate Ferrotransferrin. Apotransferrin remains tightly bound to its receptor until the complex is recycled to the cell surface, where Apotransferrin is released into the extracellular space to recruit more Fe3+ ions.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, dog and guinea pig Transferrin. Detects a band of ~76kDa by Western blot.

Type: Primary
Antigen: TRANSFERRIN
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol TF TF
Antigen name Transferrin Transferrin
Environmentally Preferable
Antigen synonyms PRO2086|Beta-1 metal-binding globulin|PRO1557|TFQTL1|Serotransferrin|Siderophilin PRO2086|Beta-1 metal-binding globulin|PRO1557|TFQTL1|Serotransferrin|Siderophilin
Antibody type Primary Primary
Clonality Polyclonal Polyclonal
Reactivity Human Human
Host Rabbit Rabbit
Isotype
Western blot Yes Yes
ImmunoPrecipitation Yes Yes
Form Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Purification Protein A affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR