Anti-BASSOON Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAM-PS003D VAM-PS003F
CA89106-616EA 443.98 CAD
CA89106-616 CA89106-618
Anti-BASSOON Mouse Monoclonal Antibody
Antibodies
Bassoon is a 420 kDa protein that localizes at the presynaptic nerve terminals and is believed to play a role in the structural and functional organization of the synaptic vesicle cycle. Bassoon is concentrated at sites opposite to postsynaptic densities in synaptic terminals and in cultured neurons, it is found to colocalize with GABA (A) and glutamate (GluR1) receptors. These observations suggested that they serve specific functions at synaptic junctions and may be involved in organization of the cytoskeleton at the site of neurotransmitter release.

  • For Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes mouse and rat Bassoon. Detects a band of ~400kDa by Western blot.

Type: Primary
Antigen: BASSOON
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR