Anti-KDEL Receptor Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAA-PT048D VAA-PT048F
CA95046-432EA 415.54 CAD
CA95046-432 CA95046-434
Anti-KDEL Receptor Mouse Monoclonal Antibody
Antibodies
The tetrapeptide KDEL, located at the carboxy-terminal sequences of luminal proteins, is a retrieval motif essential for the precise sorting of these proteins along the secretory pathway. KDEL proteins perform essential functions in the endoplasmic reticulum (ER) related to protein folding as well as assembly. The localization of chaperones and other soluble proteins to the ER is achieved by their continuous retrieval from post-ER compartments by the KDEL receptor (Erd2p), which is a membrane protein localized in the Golgi apparatus.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat KDEL Receptor. Detects a band of ~23kDa by Western blot.

Type: Primary
Antigen: KDEL Receptor
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR