Anti-ESR1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
SRA-1010F ADI-SRA-1010-F SRA-1010D ADI-SRA-1010-D
CA200063-286EA 792.01 CAD
200063-286 CA200063-286 200062-846 CA200062-846
Anti-ESR1 Mouse Monoclonal Antibody
Antibodies
The Estrogen receptors (ER), mainly ERalpha and ERbeta, are members of the nuclear steroid hormone receptor family and are composed of three domains, an N-terminal domain required for maximal transactivation function (TAF), a DNA binding domain, and a steroid binding domain. Estrogen has pleiotropic effects on the growth, development, and maintenance of diverse tissues.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and hamster estrogen receptor alpha. Detects a band of 66.3kDa by Western blot.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR