Anti-ERP72 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPS-720D SPS-720F
CA95056-564EA 346.02 CAD
CA95056-564 CA95056-566
Anti-ERP72 Rabbit Polyclonal Antibody
Antibodies
Endoplasmic reticulum protein (ERp) 72, a 72 kDa protein localized to the endoplasmic reticulum, is a member of the Protein Disulfide Isomerase (PDI) family of proteins. Together with other ER resident proteins such as BiP, GRP94, and PDI, they serve as the molecular chaperones for proper folding of newly translocated and glycolsylated proteins such as Thyroglobulin (Tg) and Human chorionic gonadotropin (hCG)-beta.

  • For Immunofluorescence, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey and rabbit ERp72. Detects a band of ~72kDa by Western blot.

Type: Primary
Antigen: ERP72
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR