Anti-HSF1 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
SPA-950D SPA-950F
CA95046-318EA 395 CAD
CA95046-318 CA95046-320
Anti-HSF1 Rat Monoclonal Antibody
Antibodies
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.

  • For Western Blot

Recognizes human, mouse, rat, monkey and rabbit HSF1. Predicted MW of ~60kDa. Detects a band of ~85kDa by Western blot.

Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR