Anti-HSF1 Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
SPA-901D SPA-901F SPA-901J
CA101410-578EA 346.02 CAD
CA101410-578 CA101410-580 CA95045-164
Anti-HSF1 Rabbit Polyclonal Antibody
Antibodies
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.
  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, EMSA
Recognizes human, mouse,More Product Information