Anti-HMOX5 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-897D SPA-897F
CA95046-314EA 346.02 CAD
CA95046-314 CA95046-316
Anti-HMOX5 Rabbit Polyclonal Antibody
Antibodies
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin. These products of the HO reaction have important physiological effects: carbon monoxide is a potent vasodilator and has been implicated to be a physiological regulator of cGMP and vascular tone; biliverdin and its product bilirubin are potent antioxidants; "free" iron increases oxidative stress and regulates the expression ofMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol HMOX4 HMOX5
Environmentally Preferable
Antigen name Heme Oxygenase 4 Heme Oxygenase 5
Antibody type Primary Primary
Clonality Polyclonal Polyclonal
Reactivity Human Human
Host Rabbit Rabbit
Isotype
Western blot Yes Yes
ELISA Yes Yes
ImmunoChemistry Yes Yes
ImmunoPrecipitation Yes Yes
ImmunoFluorescence Yes Yes
Form Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Purification Protein A affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR