Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-896D SPA-896F
CA89106-602EA 366.56 CAD
CA89106-602 CA89106-604
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
Heme Oxygenase-1 (HO-1) also known as Hsp32, is the inducible isoform of heme oxygenase that catalyzes the NADPH, oxygen, and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, ferrous iron and biliverdin which is rapidly reduced to bilirubin. These products of the HO reaction have important physiological effects: carbon monoxide is a potent vasodilator and has been implicated to be a physiological regulator of cGMP and vascular tone; biliverdin and its product bilirubin are potent antioxidants; "free" iron increases oxidative stress and regulates the expression ofMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol HMOX1
Environmentally Preferable
Antigen name Heme Oxygenase 1
Antigen synonyms Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
Antibody type Primary
Clonality Polyclonal
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
ImmunoPrecipitation Yes
Form Liquid
Purification Protein A affinity purified
Storage buffer PBS containing 50% glycerol and 0.09% sodium azide.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR