Anti-HSP90 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
SPA-835F SPA-835D
CA200062-756EA 815.28 CAD
CA200062-756 CA200063-264
Anti-HSP90 Rat Monoclonal Antibody
Antibodies
The Hsp90 family of heat shock proteins represents one of the most abundantly expressed and highly conserved families of cellular chaperones whose expression can be upregulated under conditions of cellular stress, and includes cytoplasmic (Hsp90-alpha/beta), ER (grp94), and mitochondrial (TRAP1) localized members. Structurally, Hsp90 is characterized by an N-terminal ATP-binding domain, a medial substrate-binding domain, and a C-terminal dimerization motif. Hsp90 dimers function in cooperation with cochaperones (e.g. Hsp40, Hsp70, Hop, p23) to stabilize a multitude of client protein substrates, including steroid hormone receptors, protein kinases, and transcription factors. The essential binding and hydrolysis of ATP by Hsp90 is inhibited by ansamycin drugs (e.g. geldanamycin, 17-AAG) which occupy the N-terminal Hsp90 nucleotide-binding pocket. Many Hsp90 client proteins such as erbB2/Her-2, c-raf, bcr-abl, p53, and hTERT, are members of well characterized oncogenic pathways, making Hsp90 inhibitors useful anticancer agents.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, beluga, dog, chicken, Drosophila , fish, guinea pig, hamster, monkey, mussel, opossum, pig, plant, rabbit, sheep, scallop, and Xenopus HSP90. Detects a band of ~90kDa by Western blot.

Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR