Anti-HSP60 Mouse Monoclonal Antibody
Supplier: Enzo Life Sciences
SPA-806D SPA-806F SPA-806J
CA200062-666EA 415.54 CAD
CA200062-666 CA200062-668 CA95046-188
Anti-HSP60 Mouse Monoclonal Antibody
Antibodies
Hsp60 is a member of the chaperonin family of heat shock proteins, with homologs functioning in the cytosol and mitochondria to fold nascent and aggregated proteins. Hsp60 is the eukaryotic homolog of the E. coli GroEL protein, and forms a multimeric complex in the mitochondria with Hsp10 (Cpn10) to form a large central cavity in which ATP-dependent protein folding takes place. TRiC/CCT, a eukaryotic relative of Hsp60, is expressed in the cytosol and participates in the folding of actin and tubulin substrates, but lacks any association with an Hsp10-like co-factor.