Anti-ERP57 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
SPA-725D SPA-725F
CA95046-156EA 423.44 CAD
CA95046-156 CA95046-158
Anti-ERP57 Mouse Monoclonal Antibody
ERp57 (also known as Grp58), a member of the Thioredoxin superfamily, is an ER protein with disulfide isomerase activity. It has been shown to increase after oncogenic transformation and to be a component of the MHC Class I peptide-loading complex.

  • For Immunohistochemistry, Western Blot

Recognizes human, dog, hamster, monkey and pig ERp57. Detects a band of ~58kDa by Western blot.

Type: Primary
Antigen: ERP57
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR