Anti-PTGES3 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-670D SPA-670F
CA95046-152EA 436.08 CAD
CA95046-152 CA95046-154
Anti-PTGES3 Rabbit Polyclonal Antibody
Antibodies
p23 is a ubiquitous, highly conserved co-chaperone for Hsp90 that participates in the folding of a number of cell regulatory proteins including progesterone and glucocorticoid receptors, HSF1, and telomerase. p23 is thought to modulate Hsp90 activity in the last stages of the chaperoning pathway by binding and stabilizing Hsp90 in its ATP-bound state.

  • For Western Blot

Recognizes human, mouse and rat p23. Detects a band of ~23kDa by Western blot.

Type: Primary
Antigen: PTGES3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR