Anti-CRP55 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-600D SPA-600F SPA-600J ADI-SPA-600-J ADI-SPA-600-D ADI-SPA-600-F
CA200062-592EA 346.02 CAD
CA200062-592 CA200062-594 CA95046-142 95046-142 200062-592 200062-594
Anti-CRP55 Rabbit Polyclonal Antibody
Antibodies
Calreticulin (CRT) is a multifunctional, multi-compartmental protein most abundant in the ER lumen. CRT contains the ER-retrieval sequence, KDEL, and has been best characterized as a soluble molecular chaperone of new or misfolded proteins and a Ca2+- binding protein. Both CRT and its membrane bound homolog, Calnexin (CNX) interact with proteins and glycoproteins that have monoglucosylated N-glycans. The CRT/CNX cycle promotes correct folding, inhibits aggregation of folding intermediates, blocks premature oligimerization, regulates ER degradation, and provides quality control by preventing incompletely folded glycoproteins from exiting to the Golgi complex.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, and dog calreticulin. Detects a band of ~63kDa by Western blot.

Type: Primary
Antigen: CRP55
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR