Anti-DNAJ Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-410D SPA-410F
CA200062-552EA 322.32 CAD
CA200062-552 CA200062-554
Anti-DNAJ Rabbit Polyclonal Antibody
Antibodies
DnaJ is a basic 41 kDa E. coli Heat shock protein which belongs to the molecular chaperone class of proteins. Bacterial DnaJ protein acts synergistically with the bacterial chaperone DnaK (Hsp70 homologue) and the other co-chaperone GrpE in suppressing eukaryotic and prokaryotic polypeptide aggregation, facilitating protein translocation through intracellular compartments or protein secretion, reactivating some partially aggregated enzymes, and activating pre-priming complex during initiation of DNA replication.

  • For Immunoprecipitation, Western Blot

Recognizes bacteria DnaJ. Detects a band of ~41kDa by Western blot.

Type: Primary
Antigen: DNAJ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR