Anti-CRYAB Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-226D SPA-226F
CA95046-116EA 437.66 CAD
CA95046-116 CA95046-118
Anti-CRYAB Rabbit Polyclonal Antibody
Antibodies
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye. Phosphorylation of serine residues which occurs during development and in response to stress, is intimately linked with its function. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, chicken, dog, guinea pig, hamster, monkey, pig, rabbit, scallop and Xenopus alphaB-crystallin. Detects a band of ~22kDa by Western blot.

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR