Anti-CRYAB Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
SPA-225C SPA-225E
CA200062-522EA 304.94 CAD
CA200062-522 CA200062-524
Anti-CRYAB Rabbit Polyclonal Antibody
Antibodies
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye. Phosphorylation of serine residues which occurs during development and in response to stress, is intimately linked with its function. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation.
  • For Immunoprecipitation, Western Blot
Recognizes human, mouse and bovine alphaB-crystallin.
Type: Primary
Antigen: CRYAB
More Product Information