Anti-GROES Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-210D SPA-210F
CA200062-506EA 298.62 CAD
CA200062-506 CA200063-248
Anti-GROES Rabbit Polyclonal Antibody
Antibodies
GroEL and GroES are E. coli chaperonins, which are homologs of mitochondrial chaperonins Hsp60 and Hsp10. GroEL is a double toriodal assembly of 14 identical subunits which form two heptameric rings stacked back-to-back, with a cavity at each end. GroEL and its co-chaperonin GroES facilitate protein folding with an ATP-dependent mechanism.

  • For Western Blot

Recognizes bacteria GroES. Detects a band of ~10kDa by Western blot.

Type: Primary
Antigen: GROES
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR