Anti-Ubiquitin Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-200D SPA-200F
CA200062-494EA 346.02 CAD
CA200062-494 CA200062-496
Anti-Ubiquitin Rabbit Polyclonal Antibody
Antibodies
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by the 26S complex, a multicatalytic cytosolic and nuclear protease. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

  • For Immunocytochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, fish, monkey, hamster, rabbit, guinea pig, pig, sheep, Xenopus and yeast Ubiquitin. Detects a band of ~10kDa by Western blot.

Type: Primary
Antigen: UBIQUITIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR