Anti-HSP110 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-1101E ADI-SPA-1101-E
CA95046-108EA 447.99 CAD
95046-108 CA95046-108
Anti-HSP110 Rabbit Polyclonal Antibody
Antibodies
Hsp 110 belongs to a family of large stress proteins known as the Hsp110/SSE family, the mostly overlooked relatives of the well studied Hsp70 family. Members of this group, while significantly larger and containing sequences not present in the Hsp70 family, share 30-33% amino acid identity with members of the Hsp70 family, mostly in the conserved ATP-binding domain. Presently, all identified members of the Hsp70-like sequences belong to either the Hsp70 family or the Hsp110/SSE family. Hsp110 appears to be a normal constituent of mammalian cells with highest expression in the brain, and its induction by heat shock strongly correlates with cellular thermotolerance. Hsp110 may contribute to RNA stability, and its capacity to bind peptide has made Hsp110 a promising immunoadjuvant in anti-tumor vaccine development.

  • For Western Blot

Recognizes human, mouse, rat, bovine, hamster, monkey, sheep and yeast HSP110. Detects a band of ~110kDa by Western blot.

Type: Primary
Antigen: HSP110
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR