Anti-SOD2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SOD-110F ADI-SOD-110-F SOD-110D ADI-SOD-110-D
CA95046-104EA 814 CAD
CA95046-104 95046-102 95046-104 CA95046-102
Anti-SOD2 Rabbit Polyclonal Antibody
Antibodies
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Mn SOD. Detects a band of ~25kDa by Western blot.

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR