Anti-AURKB Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-CC203D KAP-CC203F
CA101410-528EA 459.78 CAD
CA101410-528 CA101410-530
Anti-AURKB Rabbit Polyclonal Antibody
Antibodies
Aurora A, B, and C have emerged as key mitotic regulators of mammalian centrosome function, chromosome segregation and cytokinesis. Aurora B is one of several chromosomal passenger proteins involved in chromosome alignment at the metaphase plate and cytokinesis. Phosphorylation of a threonine residue in the Aurora activation loop (Thr232 in human Aurora B) is believed to be an essential regulatory mechanism in Aurora kinase activation.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse (weak), rat (weak), bovine (weak) and pig (weak) Aurora B.

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR