Anti-PKC alpha Mouse Monoclonal Antibody
Supplier: Enzo Life Sciences
KAM-PK020D KAM-PK020F
CA95045-928EA 395 CAD
CA95045-928 CA95045-930
Anti-PKC alpha Mouse Monoclonal Antibody
Antibodies
Protein Kinase C (PKC) is a large superfamily of serine/threonine kinases that mediate essential cellular signals required for activation, proliferation, differentiation and survival. There are at least ten PKC isotypes that are closely related in structure but that have distinct patterns of tissue distribution and function. The PKC isotypes can be subdivided into three classes based on primary structure and biochemical properties. These are: classical or conventional PKC isotypes (cPKC), novel PKC isotypes (nPKC) and atypical PKC isotypes (aPKC).
  • For Immunofluorescence,
More Product Information