Anti-Histone H2AX Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
KAM-CC255D KAM-CC255F
CA95045-924EA 428.18 CAD
CA95045-924 CA95045-926
Anti-Histone H2AX Mouse Monoclonal Antibody
Antibodies
The nucleosome is made up of four core histone proteins (H2A, H2B, H3 and H4) and is the primary building block of chromatin. The core histones form an octamer, which is made up of one H3-H4 tetramer and two H2A-H2B dimers. The N-terminal tail of core histones undergoes different post-transcriptional modifications including acetylation, phosphorylation, methylation and ubiquitination. These modifications occur in response to cell signal stimuli and have a direct effect on gene expression.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit and sheep Histone H2AX. Detects a band of ~16kDa by Western blot.

Type: Primary
Antigen: HISTONE H2AX
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR