Anti-TCP1A Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
CTA-123D CTA-123F
CA200061-818EA 402.9 CAD
CA200061-818 CA200063-088
Anti-TCP1A Rat Monoclonal Antibody
Antibodies
TCP-1alpha is a 60 kDa subunit of a cytosolic hetero-oligomer chaperone that is known to be involved in the folding of Actin and Tubulin. This protein is a member of the Chaperonin family, which includes E. coli GroEL, the mitochondrial Heat-shock protein Hsp60, the plastid Rubisco-subunit-binding protein, and the archaebacterial protein TF55. These Chaperonins assist the folding of proteins upon ATP hydrolysis.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, chicken, dog, hamster, plant, rabbit and sheep TCP-1alpha. Detects a band of ~60kDa by Western blot.

Type: Primary
Antigen: TCP1A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR