Anti-ACT1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
CSA-400D CSA-400F
CA95045-896EA 346.02 CAD
CA95045-896 CA95045-898
Anti-ACT1 Rabbit Polyclonal Antibody
Antibodies
Actin is one of the major proteins (~42 kDa) of the cytoskeleton. It regulates contractile potential in muscle tissues and helps to control the shape as well as the motility of non-muscle cells. Actin is expressed in all eukaryotic cells which makes it a useful loading control in Western blotting experiments. In higher eukaryotes, there are six isoforms of actin: Three alpha-actins (α-skeletal, α-cardiac, α-smooth muscle), one β-actin (β-non-muscle) and two γ-actins (γ-smooth muscle and γ-non-muscle). Actin isoforms show >90% overall sequence homology.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Actin. Detects a band of ~42kDa by Western blot.

Type: Primary
Antigen: ACT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR