Anti-AGRN Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
AGR-530D AGR-530F
CA200061-660EA 437.66 CAD
CA200061-660 CA200061-662
Anti-AGRN Mouse Monoclonal Antibody
Antibodies
Agrin is an essential extracellular matrix component which promotes clustering of nicotinic acetylcholine receptors (nAChRs) and other proteins at the neuromuscular junction during development. Agrin, MuSK, and Rapsyn are all essential components for AChR aggregation, through an unknown mechanism. The C-terminal region of agrin is released into the medium, interacts with receptors on the muscle surface and induces AChR aggregation.

  • For Immunocytochemistry, Western Blot

Recognizes mouse and rat agrin. Detects a band of ~100kDa by Western blot.

Type: Primary
Antigen: AGRN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR