Anti-BAK Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
AAP-030D AAP-030F
CA95045-878EA 415.54 CAD
CA95045-878 CA95045-880
Anti-BAK Rabbit Polyclonal Antibody
Antibodies
Bax is an appromimately 22 kDa pro-apoptotic protein belonging to the Bcl-2 family of proteins, which are characterized by the presence of at least one Bcl-2 homology (BH) domains. Bax normally resides in the cytoplasm, but following an apoptotic stimuli, Bax undergoes a conformational change and translocates to mitochondrial membranes where it inserts and mediates the release of cytochrome c. This activates caspase-3 and perpetuates the apoptotic cascade.

  • For Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, hamster, monkey and pig Bak. Detects a band of ~30kDa by Western blot.

Type: Primary
Antigen: BAK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR