Anti-PAGA Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-619-100 905-619
CA200061-462EA 531.01 CAD
200061-462 CA200061-462
Anti-PAGA Mouse Monoclonal Antibody
Antibodies
The exotoxin produced by B. anthracis is composed of three proteins: a cell-receptor binding protein known as Protective Antigen (PA), and two enzymes, Edema Factor (EF) and Lethal Factor (LF). An 83 kDa form of PA binds to a cell surface receptor and, upon proteolytic activation to a 63 kDa fragment, heptamerizes to form a membrane channel that mediates the entry of three molecules of LF or EF into the cells. PA and LF form Lethal Toxin, which is the dominant virulence factor produced by B. Anthracis and is the major cause of death in infected animals.

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes bacteria Anthrax Protective Antigen (PA).

Type: Primary
Antigen: PAGA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR