Hydrochloric acid 0.1 M

Supplier: Enzo Life Sciences

80-0080 80-0082
CA95052-780EA 105.86 CAD
CA95052-780 CA95052-782
Hydrochloric acid 0.1 M
Hydrochloric acid

Assay diluent and lysis reagent for cyclic nucleotides.

Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR