Corning® Endothelial Cell Growth Supplement, Corning
Supplier: Corning
354006 356006
CACB354006EA 235.08 CAD
CACB354006 CACB356006
Corning® Endothelial Cell Growth Supplement, Corning
Cell Culture Supplements and Additives
Endothelial Cell Growth Supplement (ECGS) is a broadly used supplement to culture a variety of cells, particularly endothelial cells.
ECGS contains various growth factors (e.g., acidic FGF or ECCF-a).
Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.