Corning® Mouse native epidermal growth factor (from male submaxillary glands)
Supplier: Corning
354001 354010
CACB354001EA 364.78 CAD
CACB354001 CACB354010
Corning® Mouse native epidermal growth factor (from male submaxillary glands)
Proteins and Peptides
Epidermal growth factor (EGF) is a low-molecular weight mitogenic protein that stimulates proliferation of a wide variety of cell types in vitro. EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.
  • Media additive
EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.
Mouse natural EGF are tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. Human recombinant EGF are shown to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. All products are filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.