Manometers, Datalogging, Sper Scientific

Supplier: Sper Scientific
840086 840086C 840098 840098C 840099 840099C
97035-994EA 735.16 CAD
97035-994 97035-996 97036-002 97036-004 97036-006 97036-008
Manometers, Datalogging, Sper Scientific
Manometers
Automatically log up to 12,000 readings with date and time stamps and download them to your computer or manually record and review 99 readings on the meter's display

Features a user friendly menu, backlit dot matrix display, auto power off and a tripod screw. This portable meter is ideal for measuring low differential pressure in clean rooms, test and balance, medical equipment, HVAC, pneumatic systems and computer peripherals. Reads gauge and differential pressure in 11 units of measure.

Certifications: N.I.S.T. traceable certificate of calibration (for certified models only).

Order Now

SPECIFICATIONS

Environmentally Preferable
Dimensions 208 x 70 x 40 mm (6.75 x 2.75 x 1.5")
Weight 255 g (9 oz.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR