Manometers, Sper Scientific
Supplier: Sper Scientific
840080 840080C 840081 840081C 840082 840082C 840083 840083C 840085 840085C
97035-974EA 1176.67 CAD
97035-974 97035-976 97035-978 97035-980 97035-982 97035-984 97035-986 97035-988 97035-990 97035-992
Manometers, Sper Scientific
Manometers
Five models are available covering a wide range.
Manometers are ideal for measuring low differential pressure in clean rooms, test and balance, medical equipment, HVAC, pneumatic systems and computer peripherals. Portable, and weighing only 5 oz (150 g) including the battery, these manometers are convenient for use in the lab, field or factory. Five models are available covering a wide range. All read gauge, and differential pressure in up to 11 units of measure and feature large backlight display, min/max, hold, and auto power off functions. The RS232 port enables communication with a computer. High accuracy of ±0.3% fs at 25ºC, repeatability: ±0.2% (max ±0.5%) FSO, and combined linearity and hysteresis: ±0.2 (Max ±1.0%) FSO.
Certifications: N.I.S.T. traceable certificate of calibration (for certified models only).
Ordering information: Manometers come ready to use with connection hose, instructions, and a 9V battery, in a hard shell carrying case. 1-year warranty.