Vacuum Meters, Sper Scientific
Supplier: Sper Scientific
840064 840064C
97035-948EA 2616.35 CAD
97035-948 97035-950
Vacuum Meters, Sper Scientific
Pumps Vacuum Gauges
Portable meter for absolute vacuum pressure measurements in hospital labs, crystal growing, solvent recovery, distillation, and rotary evaporation
Also used in HVAC, automotive and other industrial applications. Includes a detachable heavy-duty probe with ¼ PS connector for liquid, air, oil and gas. Features a large display with eight units of measure, min/max, hold, low battery indicator, auto power off and an RS232 computer interface.
Certifications: N.I.S.T. traceable certificate of compliance (for certified model only).
Ordering information: 1-year warranty. Comes ready to use in a foam-lined, hard-shell carrying case, with 9V battery and instructions.