Upchurch Scientific® Frit-In-A-Ferrule™, IDEX Health & Science
Supplier: Upchurch Scientific
P-270 P-374 P-272 P-371X P-372 P-273 P-275
53501-430EA 20.7 CAD
53501-430 53501-458 53521-016 53521-142 53521-144 53521-020 53501-454
Upchurch Scientific® Frit-In-A-Ferrule™, IDEX Health & Science
Filters Precolumn Filters and Frits
Upchurch Scientific® Frit-In-A-Ferrule™ seal and filter simultaneously by incorporating a frit into the body of a flat-bottom ferrule.
Use a Frit-In-A-Ferrule™ to filter at any point in your system where ¹/₁₆ or ¹/₈" Ext.Ø tubing is used in a flat-bottom 1/4-28, M6 or 5/16-24 connection. This product line is available in both Flangeless and Super Flangeless™ versions.
This simple yet innovative design allows one to eliminate traditional inline filters and reduce the number of additional connections in a system.
Certifications: These products are RoHS certified.
Ordering information: The Super Flangeless™ versions for ¹/₈" Ext.Ø tubing can not be used in M6 ports.