Phenolphthalein 5 g/L in diethylene glycol monoethyl ether APHA indicator for titration

Supplier: Hach
Danger

16232 16253
CA162-32EA 47.92 CAD
CA162-32 CA11016-626
Phenolphthalein 5 g/L in diethylene glycol monoethyl ether APHA indicator for titration
Phenolphthalein

pH indicator for use in titrations


Color change: colorless to pink at ~pH 8.3.

Formula: C₂₀H₁₄O₄
MDL Number: MFCD00005913
CAS Number: 77-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR