Sodium thiosulfate solution 0.0250 N stabilized

Supplier: Hach

Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfiteAntichlorSodiumthiosulphate

2409353 2409353 2409353
CA11016-454EA 116.13 CAD
CA11016-454 89231-070 104086-206
Sodium thiosulfate solution 0.0250 N stabilized
Sodium thiosulphate

For dissolved oxygen determination by Winkler titration


Stabilized sodium thiosulfate formulation.

Formula: Na₂S₂O₃
MW: 158.11 g/mol
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR