Amidosulfonic acid, powder pillow (Winkler)

Supplier: Hach
Warning

107399 2076266
CA1073-99PK 112.26 CAD
CA1073-99 CA11016-396
Amidosulfonic acid, powder pillow (Winkler)
Amidosulphonic acid

For determination of Dissolved Oxygen by the Winkler titration method


Range: 1 to >10 mg/L. Hach method 8229. Sample size: 300 mL.


Ordering information: 100/pk. Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.

Formula: H₃NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
UN: 2967
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR